Masterflex® Fitting, Natural Polypropylene, Tee, Hose Barb to Threaded Adapter, 3/4" ID x 3/4" ID x 1/2" NPT(M); 10/PK
N° de catalogue MFLX40610-83
Fournisseur: Avantor Fluid Handling
CE certificate
À propos de cet article
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
- Taille de l'ardillon/Tube Ø int. (port 2):¾"
- Type de connection:Thread to Barb
- Flow pattern:T
- Désignation:Fitting, Natural Polypropylene, Tee, Hose Barb to Threaded Adapter, ¾" ID × ¾" ID × ½" NPT(M); 10/PK
- Seq:10
- Environmentally Preferable:
- Modèle:40610-83
- Orifice dimensions:0,5625"
- Sterilisation:Ethylene oxide (ETO) or gamma
- Garantie:Out of box
- Cdt:10
- Fitting material:Polypropylene
Frequently Bought Together