Standard for osmometer
Standard for osmometer
200 mOsm calibration standard, 10×2 ml, 10
N° de catalogue ADII3MA020
Marketsource
Fournisseur:   Advanced Instruments
Standard for osmometer
N° de catalogue ADII3MA020
Fournisseur:  Advanced Instruments
Marketsource
Numéro CAS:  
Standard for osmometer
200 mOsm calibration standard, 10×2 ml, 10
Numéro CAS:  
Prix
81,00 €
Connexion Pour la disponibilité
1 * 10 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Calibration standard is used to check the perforance of osmometer and test methods.
Documents
Spécifications
Spécifications
  • Cdt:
    10
  • Désignation:
    200 mOsm calibration standard, 10×2 ml
Spécifications