Clipboards
Clipboard, A4 portrait with backclip, 240×400 mm
N° de catalogue TEKMWW0200SSCCLIP
Fournisseur: TEKNOMEK
Aller à :
Product Details & Documents
The clipboards come in a range of options.
- Brushed dull polish finish
- Spring loaded stainless steel clip
- Available in portrait or landscape
- Material: Fully welded 304-grade stainless steel
The stainless steel clipboards come in sizes A4 and A3, and can be supplied with or without a clip at the back to hang them from our hanging rails.
Spécifications
- Désignation:Clipboard, A4 portrait with backclip
- Seq:04
- l×h:240×400 mm