Mobile safety steps
Mobile safety steps, 2 steps and platform, 860×960×750 mm
N° de catalogue TEKMMST0750-2
Fournisseur: TEKNOMEK
Aller à :
Product Details & Documents
Our mobile safety steps offer optimal durability and toughness, being fully welded in 304 grade stainless steel, offering a non-slip surface for maximum safety.
- Whole structure comes with a brushed dull polish finish
- Stainless steel non slip check plate steps to ensure maximum safety and security for workers
- 3 castors- 2 swivel to rear and 1 retractable fixed at front
- Locking handle
- Widened legs
The steps castors and swivel ensure ease of manoeuvre, and the locking handle secures the steps ready for use. Different heights are available.
Spécifications
- Désignation:Mobile safety steps, 2 steps and platform
- Seq:01
- l×p×h:860×960×750 mm