Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
VWR®, Dispersing Aggregates

VWR®, Dispersing Aggregates

L'outil universel VWR® AT-5060 permet un remplacement facile du rotor dans les gros agrégats de dispersion, 0,1 kg
N° de catalogue 431-0289
Fournisseur:   VWR Collection
VWR®, Dispersing Aggregates
N° de catalogue 431-0289
Fournisseur:  VWR Collection
VWR®, Dispersing Aggregates
VWR®, Dispersing Aggregates

L'outil universel VWR® AT-5060 permet un remplacement facile du rotor dans les gros agrégats de dispersion, 0,1 kg

Prix
81,40 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

Choice of aggregates for a wide range of laboratory applications from 5 ml up to 20 L. The VWR® Aggregates solve the standard problems that are faced every day in the laboratory.

  • Easy to exchange
  • Easy to clean
  • High quality 316L stainless steel

Their universal geometry includes two rows of teeth, so they are suitable for a large spectrum of laboratory applications and are reliable and efficient.

The aggregates with Ø of 5 and 7 mm are for dispersing small amounts, these are suitable for the VWR® Homogeniser S model. Aggregates with Ø of 12 mm diameter are for dispersing small to medium volumes, these are most suitable for the VWR® Homogeniser S and M models.

Aggregates with Ø of 20 to 35 mm diameter are for dispersing medium volumes, these are most suitable for the VWR® Homogeniser M and L models.

The aggregates with Ø of 50 and 60 mm are for dispersing large volumes. These are suitable for the VWR® Homogeniser XL.

Aggregates can be used for the following applications:
– All standard dispersing tasks
– Grinding of animal and human tissue samples in a variety of fluids and volumes
– Sample preparation for the extraction and dissolution of organic materials
– Sample preparation for medical diagnostic procedures as well as DNA analysis
– Extractions of systems / substances from vegetable-derived samples
– Disruption of animal and human cells
– Deagglomeration of united cell structures
– Intensive mixing

Ordering information: Aggregate Tools not included - included with the drive units.

Spécifications
  • Désignation:
    L'outil universel VWR® AT-5060 permet un remplacement facile du rotor dans les gros agrégats de dispersion, 0,1 kg
  • Seq:
    12
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.