D(+)-Tréhalose dihydraté
Bottle, 1 kg
: Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate
- :6138-23-4
- :C₁₂H₂₂O₁₁·2H₂O
- :MFCD00071594
- :378.33 g/mol
- :97…99 °C
- :Température ambiante
- :202-739-6
- Type de conditionnement:Bottle
- Cdt:1 kg