Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
ToxinSensor™ single tests kit with standard

ToxinSensor™ single tests kit with standard

Sensitivity: 0,125 EU/ml, 40 assays, 1 KIT
Catalogus # GENSL00858-40
Leverancier:   Genscript
ToxinSensor™ single tests kit with standard
Catalogus # GENSL00858-40
Leverancier:  Genscript
CAS-nummer:  
ToxinSensor™ single tests kit with standard

Sensitivity: 0,125 EU/ml, 40 assays, 1 KIT

CAS-nummer:  
Price
282,00 €
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents

ToxinSensor™ single tests kit with standard is designed to be a qualitative In Vitro end-point endotoxin test for human and animal parenteral drugs, biological products, and medical devices.

  • High reproducibility
  • Broad detection range
  • Easy to use
  • Includes one bottle of standard

Tachypleus Amebocyte Lysate (TAL) as supplied is to be reconstituted with sample or control directly. After incubation for 1 hour, and in the presence of endotoxin, gelation occurs; in the absence of endotoxin, gelation does not occur. GenScript supply a series of ToxinSensor™ Single Tests Kits with Standard with different sensitivity (0,03 EU/ml, 0,06 EU/ml, 0,125 EU/ml and 0,25 EU/ml). When using the kit, you just need to add 200 µl of sample or control to TAL, then, wait for the required incubation time to allow for gel to form. There is no need to reconstitute the system with TAL Reagent Water before use.

Documents

Specificaties

Specificaties

  • Cdt:
    1 KIT
  • Désignation:
    Sensitivity: 0,125 EU/ml
  • Pour:
    40 assays
  • Cat. No.:
    GENSL00858-40
  • Sensibilité:
    0,125 EU/ml

Specificaties

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.