Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Human Recombinant RAP1A (from E. coli)

Human Recombinant RAP1A (from E. coli)

Catalog # ABCAAB97904-250
Marketsource
Supplier:   Abcam
Human Recombinant RAP1A (from E. coli)
Catalog # ABCAAB97904-250
Supplier:  Abcam
Marketsource
CAS Number:  
Human Recombinant RAP1A (from E. coli)
Human Recombinant RAP1A (from E. coli)
CAS Number:  
Price
2 310,00 €
Log in for availability
1 * 250 µG
Product Details & Documents

Recombinant human RAP1A protein is a Human Full Length protein, in the 1 to 181 aa range, expressed in Escherichia coli, with ≥85% purity and suitable for SDS-PAGE, MS.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Formulaire:
    Liquid
  • Activité biologique:
    Standard
  • Purity:
    ≥85%
  • Synonyme de protéine:
    KREV1|RAP1A|Ras-related protein Rap-1A|C21KG|G-22K|GTP-binding protein smg p21A|Ras-related protein Krev-1
  • Conjugaison:
    Unconjugated
  • Concentration:
    0,25 mg/ml
  • Source:
    E. coli
  • Cat. No.:
    ABCAAB97904-250
  • Conditions de stockage:
    Store at −20 °C
  • Formulation:
    pH: 8, Constituents: 40% Glycerol (glycerin, glycerine), 1,16% Sodium chloride, 0,316% Tris HCl, 0,077% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
  • Exempt de substance animale:
    N
  • Tested applications:
    MS, SDS-PAGE
  • Species:
    Human
  • Nom de la protéine/du peptide:
    RAP1A
  • Type de protéine/peptide:
    Recombinant
  • Température de transport:
    20

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.