Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Human Recombinant FGF2 (from E. coli)

Human Recombinant FGF2 (from E. coli)

N° de catalogue ABCAAB61845-1
Fournisseur:   Abcam
Human Recombinant FGF2 (from E. coli)
N° de catalogue ABCAAB61845-1
Fournisseur:  Abcam
Numéro CAS:  
Human Recombinant FGF2 (from E. coli)
Human Recombinant FGF2 (from E. coli)
Numéro CAS:  
Prix
1 560,00 €
Connexion Pour la disponibilité
1 * 1 mg
Product Details & Documents

Recombinant human FGF2 protein is a Human Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, FuncS.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Spécifications

Spécifications

  • Formulaire:
    Lyophilised
  • Activité biologique:
    Standard
  • Purity:
    >95%
  • Synonyme de protéine:
    FGFB|Fibroblast growth factor 2|FGF-2|Basic fibroblast growth factor|Heparin-binding growth factor 2|bFGF|HBGF-2
  • Conjugaison:
    Unconjugated
  • Source:
    E. coli
  • Cat. No.:
    ABCAAB61845-1
  • Conditions de stockage:
    Store at −20 °C
  • Formulation:
    Constituents: 0,164% Sodium phosphate
  • Exempt de substance animale:
    N
  • Tested applications:
    FuncS, SDS-PAGE
  • Species:
    Human
  • Nom de la protéine/du peptide:
    FGF2
  • Type de protéine/peptide:
    Recombinant
  • Température de transport:
    20

Spécifications