Safety spectacles, Rigi™
Safety spectacles, Rigi™
Rigi™ safety spectacles, Indoor/outdoor
N° de catalogue JSPA1RIG23IO
Fournisseur:   JSP
Safety spectacles, Rigi™
N° de catalogue JSPA1RIG23IO
Fournisseur:  JSP
Safety spectacles, Rigi™
Safety spectacles, Rigi™
Rigi™ safety spectacles, Indoor/outdoor
Prix
8,40 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Ultra-lightweight safety spectacle with a wide field of vision to provide optimal coverage.
  • Large field of vsion
  • Wide range of lenses available
  • Optimal coverage
Whatever the workplace environment, Rigi™ spectacles deliver the perfect combination of performance, safety, and active style. The unique close fitting design, and ultra light weight (19g) ensure lasting comfort and fit. With a wide field of vision and a design to fit all faces, wearers can achieve a comfortable customised fit throughout the day. The ergonomic fit with flat and thin flexofit temples, is compatible with any ear defender or helmet.
Certifications: Meets EN 166:2001.
Informations de commande: Available in a wide range of lens versions, including excellence coating and a unique selection of Blue Blocker lenses.
Spécifications
  • Désignation:
    Rigi™ safety spectacles
  • Seq:
    03
  • Oculaires:
    Indoor/outdoor