Sterilisation adhesive tapes
Sterilisation adhesive tapes
Neutral adhesive tapes, for crepe paper, 48 mm
Catalog # SPSA30004850
Marketsource
Supplier:   SPS
Sterilisation adhesive tapes
Catalog # SPSA30004850
Supplier:  SPS
Marketsource
Sterilisation adhesive tapes
Sterilisation adhesive tapes
Neutral adhesive tapes, for crepe paper, 48 mm
Price
Inquire For Price
Log in for availability
1 * 24 ST
Product Details & Documents
These adhesive tapes are adapted for all types of sterilisation.
  • The dispenser can be used for easier cutting tape
  • These products do not contain any latex or heavy metals
Documents
Specifications
  • Largeur:
    48 mm
  • Désignation:
    Neutral adhesive tapes, for crepe paper
  • Seq:
    06