Recombinat Protein,Human DPPIV CD26 Protein, Fc Tag,Human DPPIV, Fc Tag (DP4-H5266) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession NP001926.2).This protein carries a human IgG1 Fc tag at the N-terminus. The pr 1 * 50 µG
Catalog # ACRBDP4-H5266-50UG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together