ContactUsLinkComponent
Recombinat Protein,Human Apo-A1 Protein, His Tag,Human Apo-A1, His Tag (AP1-H5225) is expressed from human 293 cells (HEK293). It co ntains AA Arg 19 - Gln 267 (Accession NP000030.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 50 µG
N° de catalogue ACRBAP1-H5225-50UG
Fournisseur:  ACROBIOSYSTEMS
Recombinat Protein,Human Apo-A1 Protein, His Tag,Human Apo-A1, His Tag (AP1-H5225) is expressed from human 293 cells (HEK293). It co ntains AA Arg 19 - Gln 267 (Accession NP000030.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 50 µG
N° de catalogue ACRBAP1-H5225-50UG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together