Recombinat Protein,Human BMPR-IA ALK-3 Protein, Fc Tag,Human BMPR-IA, Fc Tag (AL3-H5259) is expressed from human 293 cells (HEK293) . It contains AA Gln 24 - Arg 152 (Accession CAA80257).This protein carries a human IgG1 Fc tag at the C-terminus. Th 1 * 1 mg
N° de catalogue ACRBAL3-H5259-1MG
Fournisseur: ACROBIOSYSTEMS
Frequently Bought Together