ContactUsLinkComponent
Recombinat Protein,Human Carbonic Anhydrase IX CA9 (38-414) Protein, His Tag,Human Carbonic Anhydrase IX (38-414), His Tag (CA9-H52 26) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession NP001207).This protein car 1 * 50 µG
N° de catalogue ACRBCA9-H5226-50UG
Fournisseur:  ACROBIOSYSTEMS
Recombinat Protein,Human Carbonic Anhydrase IX CA9 (38-414) Protein, His Tag,Human Carbonic Anhydrase IX (38-414), His Tag (CA9-H52 26) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession NP001207).This protein car 1 * 50 µG
N° de catalogue ACRBCA9-H5226-50UG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together