ContactUsLinkComponent
Recombinat Protein,Human IGFBP-7 Protein, His Tag,Human IGFBP-7, His Tag (IG7-H5240) is expressed from human 293 cells (HEK293). It contains AA Asp 30 - Leu 282 (Accession NP001544).This protein carries a polyhistidine tag at the N-terminus. The pro 1 * 1 mg
N° de catalogue ACRBIG7-H5240-1MG
Fournisseur:  ACROBIOSYSTEMS
Recombinat Protein,Human IGFBP-7 Protein, His Tag,Human IGFBP-7, His Tag (IG7-H5240) is expressed from human 293 cells (HEK293). It contains AA Asp 30 - Leu 282 (Accession NP001544).This protein carries a polyhistidine tag at the N-terminus. The pro 1 * 1 mg
N° de catalogue ACRBIG7-H5240-1MG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together