Mouse VEGF R3, Mouse IgG2a Fc Tag, low endotoxin (FL4-M5251) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Asp 770 (Accession P35917-1). 1 * 1 mg
N° de catalogue ACRBFL4-M5251-1MG
Fournisseur: ACROBIOSYSTEMS
Frequently Bought Together