Human ITGAVITGB6 Heterodimer Protein, His TagTag Free (IT6-H52E1) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) Gly 22 - Asn 707 (ITGB6) (Accession NP002201.1 (ITGAV) AAI21179.1 (ITGB6)). 1 * 100 µG
ACRBIT6-H52E1100UG
Frequently Bought Together