ContactUsLinkComponent
Recombinat Protein,Mouse TIM-3 HAVCR2 Protein, His Tag,Mouse TIM-3, His Tag (TI3-M52H1) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Arg 191 (Accession AAL65156.1).This protein carries a polyhistidine tag at the C-terminus. s a 1 * 1 mg
N° de catalogue ACRBTI3-M52H1-1MG
Fournisseur:  ACROBIOSYSTEMS
Recombinat Protein,Mouse TIM-3 HAVCR2 Protein, His Tag,Mouse TIM-3, His Tag (TI3-M52H1) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Arg 191 (Accession AAL65156.1).This protein carries a polyhistidine tag at the C-terminus. s a 1 * 1 mg
N° de catalogue ACRBTI3-M52H1-1MG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together