Recombinat Protein,Human Neprilysin MME CD10 Protein, Tag Free,Human Neprilysin, Tag Free (MME-H521b) is expressed from human 293 cells (HEK293). It contains AA Tyr 52 - Trp 750 (Accession NP000893.2).Human Neprilysin, with Gly-Pro at the N- terminu 1 * 1 mg
N° de catalogue ACRBMME-H521B-1MG
Fournisseur: ACROBIOSYSTEMS
Frequently Bought Together