ContactUsLinkComponent
Recombinat Protein,Human BTLA (31-150) Protein, Mouse IgG2a Fc Tag,Human BTLA (31-150), Mouse IgG2a Fc Tag (BTA-H5258) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession AAP44003.1).This protein carries a mouse Ig 1 * 1 mg
N° de catalogue ACRBBTA-H5258-1MG
Fournisseur:  ACROBIOSYSTEMS
Recombinat Protein,Human BTLA (31-150) Protein, Mouse IgG2a Fc Tag,Human BTLA (31-150), Mouse IgG2a Fc Tag (BTA-H5258) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession AAP44003.1).This protein carries a mouse Ig 1 * 1 mg
N° de catalogue ACRBBTA-H5258-1MG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together