VWR®, Accessoires
VWR®, Accessoires
Peltier unit, A-1200; controller, constant-temperature system A1200, used for UV1600PC, UV-3100PC, V3000PC, UV6300PC, cell no. 1, path length: 10 mm, temp. range: 10 ~ 90 °C [0,1 °C interval adjustable (temp. control accuracy : ±0,1 °C)]
N° de catalogue 634-1154
Fournisseur:   VWR Collection
VWR®, Accessoires
N° de catalogue 634-1154
Fournisseur:  VWR Collection
VWR®, Accessoires
VWR®, Accessoires
Peltier unit, A-1200; controller, constant-temperature system A1200, used for UV1600PC, UV-3100PC, V3000PC, UV6300PC, cell no. 1, path length: 10 mm, temp. range: 10 ~ 90 °C [0,1 °C interval adjustable (temp. control accuracy : ±0,1 °C)]
Prix
1 750,00 €
Connexion Pour la disponibilité
1 * 1 ST
Spécifications
  • Désignation:
    Peltier unit, A-1200; controller, constant-temperature system A1200, used for UV1600PC, UV-3100PC, V3000PC, UV6300PC, cell no. 1, path length: 10 mm, temp. range: 10 ~ 90 °C [0,1 °C interval adjustable (temp. control accuracy : ±0,1 °C)]
  • Seq:
    03