VWR®, Accessoires
VWR®, Accessoires
Sipper unit with temp. control, A-1000; controller (constant-temperature Peltier sipper system A1000, used for UV1600PC, UV-3100PC, V-3000PC, UV6300PC; cell no.: 1; path length: 10 mm; temp. range: 10 ~ 90 °C [0,1 °C interval adjustable]; temp. control accuracy: ±0,1 °C; injection speed: 1 ~ 8 ml/min [0,5 ml/min interval adjustable])
Catalog # 634-1153
Supplier:   VWR Collection
VWR®, Accessoires
Catalog # 634-1153
Supplier:  VWR Collection
VWR®, Accessoires
VWR®, Accessoires
Sipper unit with temp. control, A-1000; controller (constant-temperature Peltier sipper system A1000, used for UV1600PC, UV-3100PC, V-3000PC, UV6300PC; cell no.: 1; path length: 10 mm; temp. range: 10 ~ 90 °C [0,1 °C interval adjustable]; temp. control accuracy: ±0,1 °C; injection speed: 1 ~ 8 ml/min [0,5 ml/min interval adjustable])
Price
1 970,00 €
Log in for availability
1 * 1 ST
Specifications
  • Désignation:
    Sipper unit with temp. control, A-1000; controller (constant-temperature Peltier sipper system A1000, used for UV1600PC, UV-3100PC, V-3000PC, UV6300PC; cell no.: 1; path length: 10 mm; temp. range: 10 ~ 90 °C [0,1 °C interval adjustable]; temp. control accuracy: ±0,1 °C; injection speed: 1 ~ 8 ml/min [0,5 ml/min interval adjustable])
  • Seq:
    02