VWR®, Accessoires
Controller, sipper system A1100, sipper unit without peltier temp. control (used for UV1600PC, UV-3100PC, V3000PC, UV6300PC; cell no.: 1, path length: 10 mm; injection speed : 1 ~ 8 ml/min. [0,5 ml/min interval adjustable])
Catalog # 634-1152
Supplier: VWR Collection
Jump to:
Product Details & Documents
Specifications
- Désignation:Controller, sipper system A1100, sipper unit without peltier temp. control (used for UV1600PC, UV-3100PC, V3000PC, UV6300PC; cell no.: 1, path length: 10 mm; injection speed : 1 ~ 8 ml/min. [0,5 ml/min interval adjustable])
- Seq:04