TRAY STACKABLE WITH 8X30 SHELL VIALS 1 * 1 Pce
Catalog # MSPP884001
Supplier: MARKET SOURCE PART PROCESS
TRAY STACKABLE WITH 8X30 SHELL VIALS 1 * 1 Pce
Catalog # MSPP884001
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR®, Pipettes de transfert, Gamme Standard
Supplier: VWR Collection
VWR®, Gants en nitrile
Supplier: VWR Collection
VWR®, Pipettes sérologiques, gamme Standard
Supplier: VWR Collection
VWR®, Tubes PCR, 0,2 ml
Supplier: VWR Collection
Combitips® advanced Eppendorf Quality Positive Displacement Pipette Tips
Supplier: EPPENDORF
VWR®, Pipettes de transfert, High Performance
Supplier: VWR Collection
96-WELL TRAY WITH 4 X 21MM GLASS INSERTS 1 * 1 Pce
Supplier: MARKET SOURCE PART PROCESS
Ion exchange SPE, StratoSpheres™
Supplier: VARIAN
[EN]GEN II 96 WELL 4-BOLT PARALLEL SYNTH 1 * 1 Pce
Supplier: MARKET SOURCE PART PROCESS
[EN]SCOOP LAB TRANSFER PP RED 10-15MG CS 1 * 100 Pce
Supplier: TRADEWINDS DIRECT
TOP PFA SHEET FOR 96 WELL PHOTOREDOX BLOCK, FOR 8 X 30 VIALS 1 * 25 Pce
Supplier: MARKET SOURCE PART PROCESS