Labels for BradyJet J2000 Lab printer
Étiquettes pour imprimante, B-7425J, J20-235-7425J, PP, blanc, J2000-Lab, 38×25 mm, 1250 étiquettes/rouleau
N° de catalogue BRDY150022
Fournisseur: Brady
Aller à :
Product Details & Documents
These PP BradyJet J2000 Lab printer labels come in matching sizes and shapes for most sample containers including tubes, vials, cryo containers, dishes, plates and general glassware.
- Designed to stay attached and remain legible
- Resist liquid nitrogen, freezers, hot water baths and autoclaves
They can also resist common laboratory chemicals like acetone, ethanol, toluene, xylene and IPA.
Documents
Brady Jet J2000 Laboratory Colour Label Printer
Documentation produit
Spécifications
- Imprimeur:J2000-Lab
- Type:B-7425J
- Couleur:blanc
- Désignation:Étiquettes pour imprimante
- Dimensions:38×25 mm
- Matière:PP
- Seq:12
- Modèle:J20-235-7425J
- Emballage:1250 étiquettes/rouleau