Labels for BradyJet J2000 Lab printer
Étiquettes pour imprimante, B-7425J, J20-137-7425J, PP, blanc, J2000-Lab, 50×25 mm, 1500 étiquettes/rouleau
Catalog # BRDY150018
Supplier: Brady
Jump to:
Product Details & Documents
These PP BradyJet J2000 Lab printer labels come in matching sizes and shapes for most sample containers including tubes, vials, cryo containers, dishes, plates and general glassware.
- Designed to stay attached and remain legible
- Resist liquid nitrogen, freezers, hot water baths and autoclaves
They can also resist common laboratory chemicals like acetone, ethanol, toluene, xylene and IPA.
Documents
Specifications
- Imprimeur:J2000-Lab
- Type:B-7425J
- Couleur:blanc
- Désignation:Étiquettes pour imprimante
- Dimensions:50×25 mm
- Matière:PP
- Seq:16
- Modèle:J20-137-7425J
- Emballage:1500 étiquettes/rouleau