DIN adapters for electrodes
DIN adapters for electrodes
DIN 19262 adapter for LE407 / LE409 / LE420 / LE422 / LE427
N° de catalogue METT30403890
Fournisseur:   Mettler - Toledo
DIN adapters for electrodes
N° de catalogue METT30403890
Fournisseur:  Mettler - Toledo
DIN adapters for electrodes
DIN 19262 adapter for LE407 / LE409 / LE420 / LE422 / LE427
Prix
31,60 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
These adapters will allow the LE electrodes with BNC to connect to meters with DIN.
Spécifications
  • Désignation:
    DIN 19262 adapter for LE407 / LE409 / LE420 / LE422 / LE427