Tanks, rectangular, LDPE, LaboPlast®
Tanks, rectangular, LDPE, LaboPlast®
Lid, LDPE, for rectangular tanks, 400 L
Catalog # BURK3440-4001
Supplier:   BURKLE
Tanks, rectangular, LDPE, LaboPlast®
Catalog # BURK3440-4001
Supplier:  BURKLE
Tanks, rectangular, LDPE, LaboPlast®
Tanks, rectangular, LDPE, LaboPlast®
Lid, LDPE, for rectangular tanks, 400 L
Price
543,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These seamless rectangular tanks feature optimum ratio of wall thickness to filling load.
  • Linear LDPE, transparent
  • Rupture-proof
Specifications
  • Désignation:
    Lid, LDPE, for rectangular tanks, 400 L
  • Seq:
    31
Accessories
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • BURK3440-0034
    Filetage de container ¾", pour conteneurs cylindriques
    Log in for availability
    1 * 1 ST - 213,00 €