O-rings
O-ring, DN 150, Silicone
Catalog # NEUB5190-150
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
Product Details & Documents
Silicone O-rings suitable for flat flanges.
Specifications
- Désignation:O-ring, DN 150
- Matière:Silicone
- Seq:03