VWR®, DNA ladders
Mini DNA ladder, 100 − 500 bp, 100 Tests
Fournisseur: VWR Chemicals
Aller à :
Product Details & Documents
VWR® DNA ladders are convenient dsDNA ladders supplied in a loading buffer, which are ready to use on agarose and SDS DNA gels.
- Ready to use loading buffer
- For direct loading and easy visualisation
- Suitable with TBE and TAE electrophoresis systems
- Clear, distinct bands
- Blue dye front running at 100 to 300 bp at 0,5 to 1,5% agarose
Ordering information: Supplied in 0,5 ml packs for 100 lanes.
Documents
Spécifications
Spécifications
- Cdt:100 Tests
- Désignation:Mini DNA ladder, 100 − 500 bp



