Pyrogallol 98+%
Bottle, 100 g
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
- CAS Number:87-66-1
- Formula:C₆H₆O₃
- MDL Number:MFCD00002192
- Molecular Weight:126.11 g/mol
- Boiling Pt:309 °C
- Melting Pt:131…135 °C
- Density:1,45 g/cm³ (25 °C)
- Storage Temperature:Température ambiante
- EINECS:201-762-9
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Type de conditionnement:Bottle
- Cdt:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



