1-Méthylimidazole 99%
Bottle, 1 L
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: N-Methylimidazole, 1-Methyl-1H-imidazole, N-Methyl-1H-imidazole, NMI
- CAS Number:616-47-7
- Formula:C₄H₆N₂
- MDL Number:MFCD00005292
- Molecular Weight:82.11 g/mol
- Boiling Pt:198 °C
- Melting Pt:–6 °C
- Density:1,03 g/cm³ (25 °C)
- Flash Pt:92 °C
- Storage Temperature:Température ambiante
- EINECS:210-484-7
- UN:2922
- ADR:8+6.1,II
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Type de conditionnement:Bottle
- Cdt:1 L



