Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Human CD14<sup>+</sup> monocytes from peripheral blood

Human CD14+ monocytes from peripheral blood

Human peripheral blood CD14+ monocytes, 50×106 cells, Cryopreserved, Pooled, 50 Mou
N° de catalogue PB14C-3
Human CD14<sup>+</sup> monocytes from peripheral blood
N° de catalogue PB14C-3
Numéro CAS:  
Human CD14+ monocytes from peripheral blood

Human peripheral blood CD14+ monocytes, 50×106 cells, Cryopreserved, Pooled, 50 Mou

Numéro CAS:  
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 50 Mou
Product Details & Documents

Human peripheral blood CD14+ monocytes are isolated from peripheral blood mononuclear cells (PBMCs) by either positive or negative selection using immunomagnetic cell separation procedures. Peripheral blood cells are collected from normal human volunteers by direct collection or by using the COBE® Spectra or Spectra Optia® Apheresis System where cell-rich leukapheresis product is extracted using continuous flow centrifugal technology directly into the collection bag.

  • Purity >90% by flow cytometry
  • Test negative for Hepatitis B and C, HIV, HTLV, HCV/HBV, WNV, Trypanasoma cruzi, and Syphilis
  • Cryopreserved in CryoStor® CS10 media

CD14+ monocytes are responsible for phagocytosis of foreign substances in the body and are capable of killing infected host cells via antibody-mediated cellular cytotoxicity. Under the right culture conditions, monocytes can differentiate into dendritic cells, Langerhans cells, or macrophages.

Storage: Either prepare cells for long term storage in liquid nitrogen vapour phase or thaw for use. Storage in liquid phase nitrogen is NOT recommended. Short term storage of cells (less than 2 weeks) at –80 °C is acceptable, but should be minimised to ensure maximum stability. Once thawed, samples must be used immediately.

Attention: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.

Spécifications

Spécifications

  • Format de cellule:
    Cryopreserved
  • Cdt:
    50 Mou
  • Donneur unique/donneurs regroupés:
    Pooled
  • Désignation:
    Human peripheral blood CD14<sup>+</sup> monocytes, 50×10<sup>6</sup> cells

Spécifications