Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Maintenance spill berms

Maintenance spill berms

Maintenance spill berms, folded dimensions (762×406×406 mm), 76 l, 1505×905×102 mm
N° de catalogue 119-0327
Fournisseur:   JUSTRITE MANUFACTURING
Maintenance spill berms
N° de catalogue 119-0327
Fournisseur:  JUSTRITE MANUFACTURING
Maintenance spill berms
Maintenance spill berms

Maintenance spill berms, folded dimensions (762×406×406 mm), 76 l, 1505×905×102 mm

Prix
324,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

Compact and flexible spill berms quickly contain minor spills and leaks during temporary storage of drums, machine parts, batteries, and equipment. Lightweight EnGuard™I, PVC coated fabric provides broad UV and chemical resistance. Anchor berm in place using the brass grommets in each corner.

  • No assembly required; simply unfold and use
  • Encased urethane foam sidewalls offer easy roll-through access without the need for a ramp
  • Walls quickly spring back into place with no memory loss

Lightweight EnGuard™I, PVC coated fabric provides broad UV and chemical resistance. Anchor berm in place using the brass grommets in each corner. Anchor stakes not included. Order optional grates to keep products and machinery above fluid.

Certifications: Helps to comply with EPA SPCC regulations

Informations de commande: Anchor stakes not included. Order optional grates to keep products and machinery above fluid.

Spécifications
  • Capacité:
    76 l
  • Désignation:
    Maintenance spill berms, folded dimensions (762×406×406 mm)
  • Seq:
    05
  • l×p×h:
    1505×905×102 mm
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.