Connecting adapters, with parallel necks
, 18,8/26 NS, 18,8/26 NS
N° de catalogue NEUB0900-10-191919
Fournisseur: NEUBERT VOLUME GLASSWAERE
Connecting adapters, with parallel necks
N° de catalogue NEUB0900-10-191919
Fournisseur: NEUBERT VOLUME GLASSWAERE
Aller à :
Product Details & Documents
Two necked adapters made of borosilicate glass 3.3
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Spécifications
- Rodage mâle:18,8/26 NS
- Rodage femelle:18,8/26 NS
- Seq:01