Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Mouse Recombinant Noggin

Mouse Recombinant Noggin

1.000 µG
N° de catalogue STMC78061.2
Marketsource
Fournisseur:   Stemcell Technologies
Mouse Recombinant Noggin
N° de catalogue STMC78061.2
Fournisseur:  Stemcell Technologies
Marketsource
Numéro CAS:  
Mouse Recombinant Noggin
Mouse Recombinant Noggin

1.000 µG

Numéro CAS:  
Prix
3 769,00 €
Connexion Pour la disponibilité
1 * 1 EA
Product Details & Documents

Noggin binds to and antagonizes bone morphogenetic protein (BMP) ligands of the transforming growth factor beta (TGF-β) family. Noggin supports maintenance of undifferentiated human embryonic stem cells cultured in mouse embryonic fibroblast (MEF)-conditioned medium (Chaturvedi et al.), and promotes dopaminergic differentiation of embryonic stem cells and subsequent survival of dopamine neurons (Chiba et al.). Noggin is essential for development of ectodermal structures including neural tube, tooth, hair follicle, and eye, as well as patterning of mesodermal somites and skeletal structures. It also influences chondrogenesis, osteogenesis, and joint formation (Krause et al.).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Spécifications

Spécifications

  • Cdt:
    1.000 µG
  • Cat. No.:
    STMC78061.2
  • Purity:
    ≥95%
  • Synonyme de protéine:
    SYM1|SYNS1|NOG
  • Species:
    Mouse
  • Nom de la protéine/du peptide:
    Noggin
  • Type de protéine/peptide:
    Recombinant

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.