Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Human Recombinant IL-7

Human Recombinant IL-7

10 µG
N° de catalogue STMC78053.1
Marketsource
Fournisseur:   Stemcell Technologies
Human Recombinant IL-7
N° de catalogue STMC78053.1
Fournisseur:  Stemcell Technologies
Marketsource
Numéro CAS:  
Human Recombinant IL-7
Human Recombinant IL-7

10 µG

Numéro CAS:  
Prix
203,00 €
Connexion Pour la disponibilité
1 * 1 EA
Product Details & Documents

Interleukin 7 (IL-7) is a member of the Type I cytokine family that is critical for T and B cell development and survival. It is produced by non-hematopoietic cells in the thymus, lymphoid organs, and by bone marrow stromal cells (Lundström et al.). IL-7 binds to a receptor (IL-7R) composed of common gamma chain and IL-7Ra (CD127) and signals through the JAK/STAT and PI3K pathways. IL-7 regulates the survival of naïve and memory CD4+ and CD8+ T cells, γδ T cells, NK T cells, innate lymphoid cells, and T regulatory cells (Carrette and Surh). Although a deficiency in IL-7R still permits the generation of normal numbers of peripheral B cells in humans, stimulation of human B cell precursors with IL-7 could promote STAT-5-dependent proliferation and survival in vitro (Clark et al.; Corfe and Paige).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Spécifications

Spécifications

  • Cdt:
    10 µG
  • Cat. No.:
    STMC78053.1
  • Purity:
    ≥97%
  • Synonyme de protéine:
    Interleukin-7|LP-1|Lymphopoietin 1|Pre-B cell factor
  • Species:
    Human
  • Nom de la protéine/du peptide:
    IL-7
  • Type de protéine/peptide:
    Recombinant

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.