Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Mouse Recombinant IL-5

Mouse Recombinant IL-5

25 µG
Catalog # STMC78049.1
Marketsource
Mouse Recombinant IL-5
Catalog # STMC78049.1
Marketsource
CAS Number:  
Mouse Recombinant IL-5
Mouse Recombinant IL-5

25 µG

CAS Number:  
Price
190,00 €
Log in for availability
1 * 25 µG
Product Details & Documents

Interleukin 5 (IL-5) is a member of the short-chain 4-α-helical bundle subset of hematopoietic cytokines. It binds to the specific α receptor, which in turn recruits the common β receptor that initiates signaling via JAK/STAT and MAPK pathways. IL-5 is produced by Th2 cells, eosinophils, activated mast cells, and ILC2 cells. IL-5 affects the differentiation, activation, and survival of eosinophils, thus playing an important role in allergic inflammation, asthma, and parasite immunity. Stimulation of eosinophils with IL-5 leads to their activation, upregulation of CD11b expression, and inhibition of apoptosis (Shearer).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Cdt:
    25 µG
  • Cat. No.:
    STMC78049.1
  • Purity:
    ≥95%
  • Species:
    Mouse
  • Nom de la protéine/du peptide:
    IL-5
  • Type de protéine/peptide:
    Recombinant

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.