Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Human Recombinant CNTF

Human Recombinant CNTF

10 µG
N° de catalogue STMC78010
Marketsource
Fournisseur:   Stemcell Technologies
Human Recombinant CNTF
N° de catalogue STMC78010
Fournisseur:  Stemcell Technologies
Marketsource
Numéro CAS:  
Human Recombinant CNTF
Human Recombinant CNTF

10 µG

Numéro CAS:  
Prix
91,00 €
Connexion Pour la disponibilité
1 * 1 EA
Product Details & Documents

Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that belongs to the four-helix bundle cytokine family and is structurally related to interleukin 6 (IL-6), interleukin 11 (IL-11), leukemia inhibitory factor (LIF), and oncostatin M (OSM). CNTF binds to its receptor CNFTRα and induces formation of a heterodimer of the signal-transducing IL-6 receptor gp130 and LIF receptor (LIFR)-β, which triggers JAK/STAT, ERK, and the PI3K signaling cascades (Schuster et al.). CNTF plays an important role in neurogenesis and the differentiation of neural stem cells and has been suggested to possess a therapeutic role in treating neurological disorders (Ding et al.; Oppenheim et al.). CNTF has also been shown to protect rod photoreceptors from light-induced damage and to have therapeutic effects on retinal degenerative diseases caused by genetic defect or damage induced by toxins, autoantibodies, or strong light (Pernet et al.; Rhee et al.). Another therapeutic role of CNTF has been reported in protecting oligodendrocytes from death induced by apoptosis (Louis et al.). Additionally, CNTF is commonly used to differentiate human pluripotent stem cell (hPSC)-derived neural progenitor cells into astrocytes (Krencik and Zhang).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Spécifications

Spécifications

  • Cdt:
    10 µG
  • Cat. No.:
    STMC78010
  • Purity:
    ≥95%
  • Species:
    Human
  • Nom de la protéine/du peptide:
    CNTF
  • Type de protéine/peptide:
    Recombinant

Spécifications

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.