Thermobloc, SmartBlock™
Product Details & Documents
Exchangeable in just seconds. Individually sensor-controlled SmartBlocks are your solution for maintaining accurate, uniform temperatures. A wide variety of these flexible thermoblocks for all common laboratory vessels support a broad range of applications - for use with Eppendorf ThermoMixer® C and ThermoStat C.
- Optimised block design supports optimal temperature transfer to sample to achieve excellent results
- Eppendorf SmartBlock-specific calibration by integrated sensor technology maximizes temperature accuracy and homogeneity
- Insulated thermoblocks for ergonomic operation - you won't burn your fingers (PhysioCare Concept®)
- Automatic block recognition for safe handling
- Optimised block design ensures maximum temperature transfer to the sample
- Eppendorf QuickRelease technology enables fast and simple block exchange – no need for tools
Every Eppendorf SmartBlock is individually inspected in production for temperature accuracy and includes a serial-number specific certificate for documentation purpose.
Informations sur les accessoires: * Indique les SmartBlocks qui peuvent être utilisés avec un couvercle et ThermoTop. Remarque : la fréquence de mélange maximale lors de l'utilisation du couvercle avec des plaques est de 2000 min-1.
Services: Pour une installation, un IQOQ, un étalonnage accrédité selon ISO17025, un entretien ou une réparation, contactez-nous : [email protected]
Documents
EPPE5362000.035_TDS_EN
Fiche technique
Spécifications
- Désignation:SmartBlock™ DWP 500, thermoblock pour plaques Eppendorf 96/500 μl à puits profond, couvercle inclus*
- Seq:10
- Cat. No.:460-0358
- Certification environnementale:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Low Manufacturing Impact