Pads, D-Formalizer®
Pads, D-Formalizer®
D-Formalizer® Pads, 203×254 mm
N° de catalogue LEIH38007530810
Fournisseur:   LEICA HISTOLOGY
Pads, D-Formalizer®
N° de catalogue LEIH38007530810
Fournisseur:  LEICA HISTOLOGY
Pads, D-Formalizer®
Pads, D-Formalizer®
D-Formalizer® Pads, 203×254 mm
Prix
168,00 €
Connexion Pour la disponibilité
1 * 25 ST
Product Details & Documents
These D-Formalizer® pads reduces formaldehyde exposure by absorbing and neutralising any excess formalin.
  • Highly durable
  • Can be used to line shelves
  • Highly contrasting blue surface
The durable blue surface of the pads provides contrast to even small pieces of tissue. The Pads are also useful for soaking up leakage and de-odorising vapors in other applications. They can be used to line shelves which hold specimen containers that may seep formaldehyde around the lid, or to line transport boxes where leaks may occur.
Spécifications
  • Désignation:
    D-Formalizer® Pads
  • L×l:
    203×254 mm
  • Seq:
    01