Labels, energy source, electrical, Brady
Energy source labels, E-7-19x13mm-SA/100, Polyester, blanc/rouge, 19×13 mm
N° de catalogue BRDY138458
Fournisseur: Brady
Aller à :
Product Details & Documents
These electrical energy source labels are made from laminated polyester.
- Permanent acrylic adhesive
- Two different sizes available
- Helps the lockout process quicker and easier for higher safety
Used to mark your facility's points of control of control for energy sources.
Certifications: ATEX
Emballage: 100 labels per pack or 5 labels per pack.
Documents
Spécifications
- Couleur:blanc/rouge
- Désignation:Energy source labels
- Dimensions:19×13 mm
- Matière:Polyester
- Modèle:E-7-19x13mm-SA/100