Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Group lockout hasps, labelled

rouge, 76×5×180 mm
  BRDY065960
 :   Brady
Group lockout hasps, labelled
  BRDY065960
 :  Brady
Group lockout hasps, labelled
Group lockout hasps, labelled

rouge, 76×5×180 mm

 
125,00 €
   
1 * 5 ST
 

These labelled group lockout hasps are constructed from tough 5052 anodised aluminium alloy.

  • 6 padlocks can be applied
  • Include tamperproof locking tabs
  • Write-on label surface accept pencils, pen or marker
  • Pen and pencil markings can be easily erased, allowing repeated use
  • Lock hole diameter: 8 mm

 : The assorted pack contains one each of the following colours: red, silver, blue, green and gold.

 
  • Couleur:
    rouge
  • Seq:
    04
  • l×p×h:
    76×5×180 mm