Sample cones for PerkinElmer
Sampler cone, 1,1 mm orifice, for ELAN 9000/6x00/DRC, not compatible with NexION, Nickel
N° de catalogue HEWL8003-0573
Fournisseur: Agilent
Aller à :
Product Details & Documents
These sample and skimmer cones are equivalent to those from PerkinElmer ICP-MS for complete confidence.
- Guaranteed to be compatible with PerkinElmer instruments
- Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
- Parts qualified on a genuine PerkinElmer system
- Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
Large-orifice sampler and skimmer cones provide superior long-term stability and resist clogging, allowing analysis under both high and low sample-uptake conditions. Nickel is a very rugged, long-lasting material.
Spécifications
- Désignation:Sampler cone, 1,1 mm orifice, for ELAN 9000/6x00/DRC, not compatible with NexION
- Matière:Nickel
- Seq:02
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.