Sample cones for PerkinElmer
Sample cones for PerkinElmer
Sampler cone, 1,1 mm orifice, for ELAN 9000/6x00/DRC, not compatible with NexION, Nickel
N° de catalogue HEWL8003-0573
Fournisseur:   Agilent
Sample cones for PerkinElmer
N° de catalogue HEWL8003-0573
Fournisseur:  Agilent
Sample cones for PerkinElmer
Sampler cone, 1,1 mm orifice, for ELAN 9000/6x00/DRC, not compatible with NexION, Nickel
Prix
604,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
These sample and skimmer cones are equivalent to those from PerkinElmer ICP-MS for complete confidence.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
Large-orifice sampler and skimmer cones provide superior long-term stability and resist clogging, allowing analysis under both high and low sample-uptake conditions. Nickel is a very rugged, long-lasting material.
Spécifications
  • Désignation:
    Sampler cone, 1,1 mm orifice, for ELAN 9000/6x00/DRC, not compatible with NexION
  • Matière:
    Nickel
  • Seq:
    02