Vydac 238TP C18, HPLC columns
Analytique, C18, Acier inoxydable, 300 Å, 5 µm, 4,60 mm, 250 mm, Silica
Catalog # 238TP54
Supplier: Avantor
Jump to:
Product Details & Documents
Vydac TP is the first range of wide pore media phases developed in the Vydac range and it is the industry's standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
These first generation monomeric C18 phase columns are recommended for the same applications as 218TP but offers alternative C18 selectivity.
* Use with holder 80101/N and column coupler HI-081.
Vydac is a registered trademark of W.R.Grace and Co.
Specifications
- Type de colonne:Analytique
- Ø int.:4,60 mm
- Granulométrie:5 µm
- Taille de pore:300 Å
- Longueur:250 mm
- Seq:18
- Matériau de colonne:Acier inoxydable
- Greffage:C18
- Matériau de remplissage:Silica
- Cat. No.:238TP54
- Certification environnementale:ISO 14001:2015 (530840 UM15)
- Code USP:L1
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Product Information
- Catalog #DésignationAvailabilityPrice
- 80101/NPorte-cartouche All-Guard pour colonnes au diamètre interne compris entre 2,1 à 4,6 mmLog in for availability1 * 1 ST - 232,00 €
Product Information
- Catalog #DésignationAvailabilityPrice



