Vydac 214TP C4, HPLC columns
Préparative, Vydac 214TP, Acier inoxydable, 300 Å, 10 - 15 µm, 22,00 mm, 250 mm, Silica
N° de catalogue 214TP101522
Fournisseur: Avantor
Aller à :
Product Details & Documents
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
214TP C4 is a first generation C4 phase and is recommended for the analysis of glycoproteins, haemoglobin variants, histones, insulin variants and membrane proteins.
Vydac is a registered trademark of W.R.Grace and Co.
Spécifications
- Type de colonne:Préparative
- Ø int.:22,00 mm
- Granulométrie:10 - 15 µm
- Taille de pore:300 Å
- Longueur:250 mm
- Seq:42
- Matériau de colonne:Acier inoxydable
- Greffage:Vydac 214TP
- Matériau de remplissage:Silica
- Cat. No.:214TP101522
- End-capping:Oui
- Code USP:L26
- Charge de carbone:3%



