Vydac 214TP C4, HPLC columns
Analytique, Vydac 214TP, Acier inoxydable, 300 Å, 5 µm, 2,10 mm, 50 mm, Silica
N° de catalogue 214TP5205
Fournisseur: Avantor
Vydac 214TP C4, HPLC columns
N° de catalogue 214TP5205
Fournisseur: Avantor
Aller à :
Product Details & Documents
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
214TP C4 is a first generation C4 phase and is recommended for the analysis of glycoproteins, haemoglobin variants, histones, insulin variants and membrane proteins.
Vydac is a registered trademark of W.R.Grace and Co.
Documents
Spécifications
- Type de colonne:Analytique
- Ø int.:2,10 mm
- Granulométrie:5 µm
- Taille de pore:300 Å
- Longueur:50 mm
- Seq:16
- Matériau de colonne:Acier inoxydable
- Greffage:Vydac 214TP
- Matériau de remplissage:Silica
- Cat. No.:214TP5205
- End-capping:Oui
- Certification environnementale:ISO 14001:2015 (530840 UM15)
- Code USP:L26
- Environmentally Preferable:Low Manufacturing Impact
- Charge de carbone:3%
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



