Vydac 214TP C4, HPLC columns
Guard cartridge for 10 – 22 mm ID columns (use with holder GCH10 and coupler HI-081), Cartouche de garde, Vydac 214TP, Acier inoxydable, 5 µm, Silica
Catalog # 214GCC510
Supplier: Avantor
Jump to:
Product Details & Documents
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
214TP C4 is a first generation C4 phase and is recommended for the analysis of glycoproteins, haemoglobin variants, histones, insulin variants and membrane proteins.
Vydac is a registered trademark of W.R.Grace and Co.
Documents
Specifications
- Type de colonne:Cartouche de garde
- Granulométrie:5 µm
- Désignation:Guard cartridge for 10 – 22 mm ID columns (use with holder GCH10 and coupler HI-081)
- Seq:54
- Matériau de colonne:Acier inoxydable
- Greffage:Vydac 214TP
- Matériau de remplissage:Silica
- Cat. No.:214GCC510
- Certification environnementale:ISO 14001:2015 (530840 UM15)
- Environmentally Preferable:Low Manufacturing Impact
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



