Vydac 208TP C8, HPLC columns
Guard kit for 10 – 22 mm ID columns, includes 1× guard cartridge, holder and coupler, Cartouche de garde, C8, Acier inoxydable, 300 Å, 12 µm, Silica
Catalog # 208FSK1210
Supplier: Avantor
Jump to:
Product Details & Documents
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
208TP C8 is less hydrophobic than C18TP and is recommended for the analysis of peptides, 10 – 20K MW.
Vydac is a registered trademark of W.R.Grace and Co.
Specifications
- Type de colonne:Cartouche de garde
- Granulométrie:12 µm
- Taille de pore:300 Å
- Désignation:Guard kit for 10 – 22 mm ID columns, includes 1× guard cartridge, holder and coupler
- Seq:31
- Matériau de colonne:Acier inoxydable
- Greffage:C8
- Matériau de remplissage:Silica
- Cat. No.:208FSK1210
- Certification environnementale:ISO 14001:2015 (530840 UM15)
- Environmentally Preferable:Low Manufacturing Impact
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



